missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Antibiotics: Actinomycin D

Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis

$121.66 - $666.34

Specifications

Content And Storage 4°C
Form Powder
Product Type Antibiotic
Sterility Non-sterile
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19452505
View Documents
MP Biomedicals™
0219452505
5 mg
Each for $159.13
Only null left
 
ICN19452510
View Documents
MP Biomedicals™
0219452510
10 mg
Each for $304.92
Only null left
 
ICN19452502
View Documents
MP Biomedicals™
0219452502
2 mg
Each for $121.66
Only null left
 
ICN19452525
View Documents
MP Biomedicals™
0219452525
25 mg
Each for $666.34
Only null left
 
Description

Description

  • Formate: Powder, Non-sterile

All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.

Specifications

Specifications

4°C
Powder
Antibiotic
Non-sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'