Learn More
Antibiotic A23187, 98%
An antibiotic and calcium ionophore | CAS: 52665-69-7 | C29H37N3O6 | 523.63 g/mol
Supplier: Thermo Scientific Chemicals J63020MA
Description
A23187 (Mixed Calcium-Magnesium Salt) is used in cell activation experiments when calcium dose-response data are not required. Ca-Mg ratio ∼ 1:1. it is widely used as a compound in cell activation. It is used as a highly selective Ca2+ ionophore. it is used in studies of intracellular Ca2+ mediated events. A23187 has been observed to induce cell activation, differentiation or proliferation. In vitro experimentation has shown A23187 to boost ROS generation and platelet fragmentation. A23187 is a widely used ionophore. It has the ability to increase the intracellular concentrations of cations and is widely used to increase the intracellular levels of calcium. Visualization of Synaptic Ca2+ Calmodulin-Dependent Protein Kinase II Activity in Living Neurons.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA23187 (Mixed Calcium-Magnesium Salt) is used in cell activation experiments when calcium dose-response data are not required. Ca-Mg ratio ∼ 1:1. it is widely used as a compound in cell activation. It is used as a highly selective Ca2+ ionophore. it is used in studies of intracellular Ca2+ mediated events. A23187 has been observed to induce cell activation, differentiation or proliferation. In vitro experimentation has shown A23187 to boost ROS generation and platelet fragmentation. A23187 is a widely used ionophore. It has the ability to increase the intracellular concentrations of cations and is widely used to increase the intracellular levels of calcium. Visualization of Synaptic Ca2+ Calmodulin-Dependent Protein Kinase II Activity in Living Neurons.
Solubility
Soluble in DMSO with warming or sonication. Also soluble in ethanol or ethyl acetate
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Specifications
| Antibiotic A23187 | |
| Powder | |
| White to Yellow | |
| C29H37N3O6 | |
| 1096801 | |
| Calcimycin; Calcium ionophore A23187 | |
| HIYAVKIYRIFSCZ-CVXKHCKVSA-N | |
| 5-(methylamino)-2-{[(2S,3S,8S,9R,11R)-3,9,11-trimethyl-8-[1-oxo-1-(1H-pyrrol-2-yl)propan-2-yl]-1,7-dioxaspiro[5.5]undecan-2-yl]methyl}-1,3-benzoxazole-4-carboxylic acid | |
| 97043288 | |
| 98% |
| Antibiotic A23187 | |
| 52665-69-7 | |
| 10 mg | |
| MFCD00151202,MFCD00151202 | |
| 14,1639 | |
| Soluble in DMSO with warming or sonication; Also soluble in ethanol or ethyl acetate | |
| CNC1=CC=C2OC(C[C@@H]3OC4(CC[C@@H]3C)O[C@H](C(C)C(=O)C3=CC=CN3)[C@H](C)C[C@H]4C)=NC2=C1C(O)=O | |
| 523.63 | |
| 523.62 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 258-084-1
RTECSNumber : DM4676000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only