missing translation for 'onlineSavingsMsg'
Learn More
Learn More
zinc finger protein 500, Mouse, Polyclonal Antibody, Abnova™
Description
Sequence: QRVHTGEKPYPCPECGKRFSQSSSLVIHRRTHSGERPYACTQCGKRFNNSSHFSAHRRTHTGEKPYTCPACGRGFRRGTDLHKHQRTHMGAGSLPTLQPVAPGGPGAKA
Specifications
Specifications
| Antigen | zinc finger protein 500 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant ZNF500. |
| Formulation | 50% glycerol |
| Gene | ZNF500 |
| Gene Accession No. | NM_021646 |
| Gene Alias | ZKSCAN18 |
| Gene Symbols | ZNF500 |
| Show More |