missing translation for 'onlineSavingsMsg'
Learn More

WNK3 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 072262MI
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
072262MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 072262MI Supplier MilliporeSigma Supplier No. 072262

Rabbit Polyclonal Antibody

This WNK3 antibody is validated for use in western blotting for the detection of the WNK3 protein.

Specifications

Antigen WNK3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols WNK3; PRKWNK3
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human WNK3.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001002838
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only