missing translation for 'onlineSavingsMsg'
Learn More

VGF Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABN652MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN652MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN652MI Supplier MilliporeSigma Supplier No. ABN652
Only null left

Rabbit Polyclonal Antibody

Anti-VGF Antibody is an antibody against VGF for use in western blotting, immunohistochemistry and immunofluorescence.

Specifications

Antigen VGF
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Gene Symbols VGF
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide corresponding to the C-terminus of human VGF.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only