missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vesicular Acetylcholine Transporter (VAChT) Goat anti-Rat, Mouse, Polyclonal, MilliporeSigma™
Goat Polyclonal Antibody
Supplier: MilliporeSigma ABN100
Description
Anti-Vesicular Acetylcholine Transporter (VAChT) Antibody detects level of Vesicular Acetylcholine Transporter (VAChT) and has been published and validated for use in immunohistochemistry, ELISA.Specifications
| Vesicular Acetylcholine Transporter (VAChT) | |
| Polyclonal | |
| Purified goat polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| Goat | |
| Affinity Purified | |
| Primary | |
| Mouse, Rat |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Slc18a3; Vacht; Vat | |
| KLH-conjugated linear peptide corresponding to the C-terminus of rat VAChT. | |
| 100 μg | |
| NP_113851 | |
| 2-8°C for 12 months |
For Research Use Only