Learn More
VEGF Mouse anti-Human, Mouse, Rat, Rabbit, Clone: JH, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma 05443
Catalog No. 05-443-MI
Only null left
Explore available promotions
Description
Description
This Anti-VEGF Antibody, clone JH is validated for use in immunohistochemistry (paraffin) for the detection of VEGF.
Specifications
Specifications
VEGF
Monoclonal
Unconjugated
NM_003376
Mouse
Protein G chromatography
Primary
Stable for 1 year at from date of receipt.
IgG1 κ
Immunohistochemistry (Paraffin)
JH
Purified mouse monoclonal IgG1k in buffer containing 10mM PBS, pH 7.4.
VEGFA, VEGF-A, VEGF, VPF, MGC70609
Recombinant human VEGF121. Clone JH121
100 μg
Human, Mouse, Rabbit, Rat
Purified
Product Suggestions
Product Suggestions