missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAC14, Mouse, Clone: 3B2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant VAC14.
Supplier: Abnova Corporation H00055697M03
Description
Phosphatidylinositol 3,5-bisphosphate (PI(3,5)P2) is a low-abundance signaling molecule. A regulatory complex made up of VAC14 and FIG4 (MIM 609390) control synthesis of PI(3,5)P2 by activating PI(3)P kinase, FAB1 (PIP5K3; MIM 609414). The VAC14/FIG4 complex also functions in the breakdown of PI(3,5)P2 (Zhang et al., 2007 [PubMed 17956977]).[supplied by OMIM
Sequence: SHRLQCVPNPELLQTEDSLKAAPKSQKADSPSIDYAELLQHFEKVQNKHLEVRHQRSGRGDHLDRRVVLSpecifications
| VAC14 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_018052 | |
| VAC14 | |
| VAC14 (NP_060522.3, 714 a.a. ∼ 782 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3B2 | |
| Mouse monoclonal antibody raised against a partial recombinant VAC14. | |
| VAC14 | |
| ArPIKfyve/FLJ10305/FLJ36622/FLJ46582/MGC149815/MGC149816/TAX1BP2/TRX | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55697 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |