missing translation for 'onlineSavingsMsg'
Learn More

UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017078
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89017078 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89017078 Supplier Abnova Corporation Supplier No. H00051118B01P
Only null left

Mouse polyclonal antibody raised against a full-length human UTP11L protein.

Sequence: MAAAFRKAAKSRQREHRERSQPGFRKHLGLLEKKKDYKLRADDYRKKQEYLKALRKKALEKNPDEFYYKMTRVKLQDGVHIIKETKEEVTPEQLKLMRTQDVKYIEMKRVAEAKKIERLKSELHLLDFQGKQQNKHVFFFDTKKEVEQFDVATHLQTAPELVDRVFNRPRIETLQKEKVKGVTNQTGLKRIAKERQKQYNCLTQRIEREKKLFVIAQKIQTRKDLMDKTQKVKVKKETVNSPAIYKFQSRRKR

Specifications

Antigen UTP11-like, U3 small nucleolar ribonucleoprotein, (yeast)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human UTP11L protein.
Formulation PBS with no preservative; pH 7.4
Gene UTP11L
Gene Accession No. BC005182.1
Gene Alias CGI-94/CGI94
Gene Symbols UTP11L
Host Species Mouse
Immunogen UTP11L (AAH05182.1, 1 a.a. ∼ 253 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 51118
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less