missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uncoupling Protein 2 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3226
Description
Detect Uncoupling Protein 2 using this Anti-Uncoupling Protein 2 Antibody validated for use in ELISA and western blotting.Specifications
| Uncoupling Protein 2 | |
| Polyclonal | |
| Affinity purified immunoglobulin. Liquid in 100mM Tris, pH 7.5 with 0.2% BSA as stabilizer and 0.05% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human |
| ELISA, Western Blot | |
| Unconjugated | |
| UCP2; UCPH; SLC25A8 | |
| 13 aa peptide sequence near the 3rd and 4th transmembrane domain of human UCP2 (Fleury et al. 1997; Gimeon et al. 1997). The peptide has no significant homology with UCP1, UCP3 or UCP4 (Boss et al. 1997). | |
| 50 μg | |
| NM_003355.2 | |
| −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only