missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-Tubulin Mouse anti-Rat, Mouse, Pig, Bovine, Avian, Guinea Pig, Human, Clone: DM1A, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05829
Description
Anti-α-Tubulin Antibody, clone DM1A is an antibody against α-Tubulin for use in immunocytochemistry, immunofluorescence and western blotting.Specifications
| α-Tubulin | |
| Monoclonal | |
| Unconjugated | |
| NP_006000 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| DM1A | |
| Purified mouse monoclonal IgG1 in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide and 30% glycerol. | |
| TUBA1A, hum-a-tub2, FLJ25113, B-ALPHA-1, TUBA3, hum-a-tub1 | |
| Purified human brain α-Tubulin. The epitope recognized by this antibody has been mapped to amino acids 426-450 (AALEKDYEEVGVDSVEGEGEEEGEE). | |
| 200 μg | |
| Avian, Bovine, Guinea Pig, Human, Mouse, Pig, Rat | |
| Purified |
For Research Use Only