Learn More
Tubulin Rabbit anti-Human, Mouse, Rat, Mammals, Green Plants, Sea Urchin, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma AB3201
Catalog No. AB3201MI
Only null left
Description
Description
Anti-Tubulin Antibody, Detyrosinated is an antibody against Tubulin for use in immunocytochemistry, immunohistochemistry (paraffin) and western blotting.
Specifications
Specifications
Tubulin
Polyclonal
Purified rabbit polyclonal in buffer containing liquid PBS.
Rabbit
Affinity Purified
Primary
Human, Mammalia, Mouse, Plant, Rat, Sea urchin
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TUBB3; TUBB4; MC1R; beta-4
Peptide corresponding to the C-terminal of Glu tubulin.
50 μg
NM_006086.2
-20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles
Product Suggestions
Product Suggestions