missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB135
Description
Anti-Tryptophan Antibody is an antibody against Tryptophan for use in immunohistochemistry.Specifications
| Tryptophan | |
| Polyclonal | |
| Rabbit antiserum. Liquid with 0.05% sodium azide. | |
| Tryptophan-gluteraldehyde-Poly-lysine | |
| Primary | |
| −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| 100 μL | |
| Rat | |
| Serum |
For Research Use Only