Learn More
Trimethyl-Histone H4 (Lys20) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma 07463S
 View more versions of this product
Catalog No. 07463SMI
Only null left
Description
Description
Anti-trimethyl-Histone H4 (Lys20) Antibody is a rabbit polyclonal antibody for detection of trimethyl-Histone H4 (Lys20) also known as H4K20me3, Histone H4 (tri methyl K20) and has been validated in immunoprecipitation, western blot, Chimmunoprecipitation, DB.
Specifications
Chemical Identifiers
trimethyl Histone H4 (Lys20)
Polyclonal
Rabbit
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
HIST1H4A; H4/J; HIST1H4F; HIST1H4L; H4FN; H4FA; H4FH; HIST1H4H; H4F2; H4FM; H4/A; H4FK; H4FG; HIST2H4; H4FB; H4/K; HIST1H4I; H4/N; HIST1H4B; H4FE; H4/M; H4/a; HIST1H4E; HIST4H4; HIST2H4A; H4FC; H4FI; H4/H; HIST1H4C; H4/G; HIST1H4K; H4FJ; H4FD; H4/I; H4/B; H4/C; H4/D; HIST1H4J; HIST1H4D; H4/E
Primary
2-8°C for 12 months
ChIP Assay, Dot Blot, Immunoprecipitation, Western Blot
Unconjugated
Human, Mouse, Rat
NP_001029249
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4 trimethylated at Lys20.
Affinity Purified
Specifications
trimethyl Histone H4 (Lys20)
Polyclonal
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Rabbit
Affinity Purified
Primary
Human, Mouse, Rat
ChIP Assay, Dot Blot, Immunoprecipitation, Western Blot
Unconjugated
HIST1H4A; H4/J; HIST1H4F; HIST1H4L; H4FN; H4FA; H4FH; HIST1H4H; H4F2; H4FM; H4/A; H4FK; H4FG; HIST2H4; H4FB; H4/K; HIST1H4I; H4/N; HIST1H4B; H4FE; H4/M; H4/a; HIST1H4E; HIST4H4; HIST2H4A; H4FC; H4FI; H4/H; HIST1H4C; H4/G; HIST1H4K; H4FJ; H4FD; H4/I; H4/B; H4/C; H4/D; HIST1H4J; HIST1H4D; H4/E
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4 trimethylated at Lys20.
10 μg
NP_001029249
2-8°C for 12 months
Product Suggestions
Product Suggestions