missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl-Histone H3 (Lys9) Mouse anti-Human, Mouse, Clone: 6F12-H4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051242S
Description
Use Anti-trimethyl-Histone H3 (Lys9) Antibody, clone 6F12-H4 (mouse monoclonal antibody) validated in Chimmunoprecipitation, PIA, immunofluoresence, DB, ChIP-seq, western blotting to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).Specifications
| trimethyl-Histone H3 (Lys9) | |
| Monoclonal | |
| Unconjugated | |
| NM_003493 | |
| Mouse | |
| Purified | |
| Primary | |
| 2-8°C for one year. Avoid freezing, which can damage IgG and affect product performance. | |
| IgG1 κ |
| ChIP Assay, Dot Blot, Immunofluorescence, Inhibition Assays, Western Blot | |
| 6F12-H4 | |
| Purified antibody supplied at 1mg/mL in 0.1MTris-glycine, pH 7.4, 150mM NaCl, 0.05% NaN3. | |
| H3.4, H3/g, H3/t, H3FT, H3t, MGC126886, OTTHUMP00000037945 | |
| KLH-conjugated, branched synthetic peptide containing the sequence (QTARme3K- STGGKA)2-KC, in which me3K corresponds to trimethyl lysine 9 of human histone H3. Data suggests this antibody may require the presence of the glutamine residue in position 5 fo | |
| 20 μL | |
| Human, Mouse | |
| Purified |
For Research Use Only