missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Histone H3 (Lys27) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE44SMI
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE44SMI 10 μg
ABE44MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE44SMI Supplier MilliporeSigma Supplier No. ABE44S
Only null left

Rabbit Polyclonal Antibody

Anti-trimethyl Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated on lysine 27 (H3K73me). This purified Ab is specificity verified by dot blot (DB) and validated in western blot, immunocytochemistry, PIA. Small pack size is ideal for screening applications.

Specifications

Antigen trimethyl Histone H3 (Lys27)
Applications Dot Blot, Immunocytochemistry, Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Host Species Rabbit
Immunogen Branch peptide corresponding to human Histone H3 methylated at Lys27.
Purification Method Affinity Purified
Quantity 10 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_003484.1
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only