Learn More
TRA-2-49 Mouse anti-Human, FITC, Clone: TRA-2-49/6E, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma FCMAB133F
Catalog No. FCMAB133FMI
Only null left
Description
Description
Detect TRA-2-49 using this Milli-Mark™ Anti-TRA-2-49 Antibody, clone TRA-2-49/6E, FITC Conjugate validated for use in flow cytometry.
Specifications
Specifications
TRA-2-49
Monoclonal
FITC
TRA-2-49
2102Ep human Embryonal Carcinoma Cells
100 Tests
Human
Purified
Flow Cytometry
TRA-2-49/6E
Purified mouse monoclonal IgG1 conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Mouse
Protein A purfied
Primary
2-8°C in undiluted aliquots for up to 6 months. Protect from light.
IgG1
Product Suggestions
Product Suggestions