missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53RK binding protein, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TPRKB protein.
Supplier: Abnova Corporation H00051002B01P
Description
Sequence: MQLTHQLDLFPECRVTLLLFKDVKNAGDLRRKAMEGTIDGSLINPTVIVDPFQILVAANKAVHLYKLGKMKTRTLSTEIIFNLSPNNNISEALKKFGISANDTSILIVYIEEGEKQINQEYLISQVEGHQVSLKNLPEIMNITEVKKIYKLSSQEESIGTLLDAIICRMSTKDVLSpecifications
| TP53RK binding protein | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human TPRKB protein. | |
| TPRKB | |
| CGI-121 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 51002 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_016058.1 | |
| TPRKB | |
| TPRKB (NP_057142.1, 1 a.a. ∼ 175 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |