missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiazide-Sensitive NaCl Cotransporter Rabbit anti-Human, Mouse, Rat, Rabbit, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3553
Description
This Anti-Thiazide-Sensitive NaCl Cotransporter Antibody is validated for use in immunohistochemistry, western blotting for the detection of Thiazide-Sensitive NaCl Cotransporter.Specifications
| Thiazide-Sensitive NaCl Cotransporter | |
| Polyclonal | |
| Affinity Purified immunoglobulin in PBS containing no preservatives. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rabbit, Rat |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| SLC12A3; NCCT; TSC | |
| Synthetic peptide from the N-terminal of human/rat/mouse NCC. | |
| 50 μL | |
| NM_000339.1 | |
| −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only