missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAL-1 Mouse anti-Human, Clone: BTL73, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04123
Description
Use Anti-TAL-1 Antibody, clone BTL73 (Mouse Monoclonal Antibody) validated in western blot, immunohistochemistry to detect TAL-1 also known as SCL, T-cell acute lymphocytic leukemia 1, TCL5, TAL-1, Stem cell protein.Specifications
| TAL-1 | |
| Monoclonal | |
| Unconjugated | |
| NP_003180 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. |
| Immunohistochemistry, Western Blot | |
| BTL73 | |
| Purified mouse monoclonal IgG1k in buffer containing PBS and 0.05% sodium azide. | |
| TAL1, SCL, TCL5 | |
| Recombinant GST-TAL1 (K Pulford, et al., 1995, Blood, 85: 675 - 684) | |
| 100 μg | |
| Human | |
| IgG1 κ |
For Research Use Only