missing translation for 'onlineSavingsMsg'
Learn More

Synaptopodin Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABN481MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN481MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN481MI Supplier MilliporeSigma Supplier No. ABN481
Only null left

Rabbit Polyclonal Antibody

Detect Synaptopodin using this rabbit polyclonal antibody, Anti-Synaptopodin Antibody validated for use in western blotting and immunohistochemistry.

Specifications

Antigen Synaptopodin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.
Gene Symbols SYNPO
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human SYNPO.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only