missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ anti-Synapsin I, Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ 57477850UL
Description
Specifically detects Synapsin I Clone: in Bovine, Human, Mouse, Rat samples, and it is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Immuno Blotting
Synapsin I is a neuron specific protein that serves as an excellent marker for synaptic terminals. Can be used to estimate synaptic density and synaptogenesis.Specifications
| Synapsin I | |
| Polyclonal | |
| Undiluted serum. | |
| purified, bovine brain synapsin I | |
| RUO | |
| Recognizes the synapsin I protein in rat brain. | |
| −20°C | |
| IgG |
| ELISA, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Undiluted |
For Research Use Only