missing translation for 'onlineSavingsMsg'
Learn More

Stem Cell Factor Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB1498PMI
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1498PMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1498PMI Supplier MilliporeSigma Supplier No. AB1498P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Detect Stem Cell Factor using this Anti-Stem Cell Factor Antibody validated for use in ELISA, western blot, NEUT.

Specifications

Antigen Stem Cell Factor
Applications ELISA, Neutralization, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Lyophilized from 0.5X PBS pH 7.2. Reconstitute with sterile distilled water.
Gene Symbols KITLG; SCF; MGF; Kitl; DKFZp686F2250; SF; KL-1
Host Species Rabbit
Immunogen Highly pure (≥98%) recombinant mouse SCF
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) NM_000899.3; NM_003994.4
Target Species Mouse
Content And Storage Lyophilized: −20°C for up to 12 months. Reconstituted: 2-4°C in undiluted aliquots for up to 6 weeks
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only