Learn More
KNF™ FLEXPAK Anti-static Flat Bags
Clear and clean polyethylene bags with a 4 mil thickness.
Supplier:  KNF™ FLEXPAK 020507529
 View more versions of this product
Catalog No. 17020295
Description
Description
  • Clear, anti-static plastic
  • Flat bottom
Specifications
Chemical Identifiers
Not Autoclavable
Clear
4 mil
Level 200 Clean
Polyethylene
0.1016 mm
Specifications
Not Autoclavable
Clear
4 mil
182.9 cm
0.1016 mm
Level 200 Clean
Polyethylene
72 x 72 in.
182.9 cm
Product Suggestions
Product Suggestions