missing translation for 'onlineSavingsMsg'
Learn More
Learn More
small proline-rich protein 3, Mouse, Clone: 4A12, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SPRR3.
Supplier: Abnova Corporation H00006707M01
Description
Sequence: MSSYQQKQTFTPPPQLQQQQVKQPSQPPPQEIFVPTTKEPCHSKVPQPGNTKIPEPGCTKVPEPGCTKVPEPGCTKVPEPGCTKVPEPGCTKVPEPGYTKVPEPGSIKVPDQGFIKFPEPGAIKVPEQGYTKVPVPGYTKVPEPCPSTVTPGPAQQKTKQKSpecifications
| small proline-rich protein 3 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC017802 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 6707 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 4A12 | |
| Mouse monoclonal antibody raised against a full length recombinant SPRR3. | |
| SPRR3 | |
| SPRR3 | |
| SPRR3 (AAH17802, 1 a.a. ∼ 161 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |