missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium/Iodine Symporter Mouse anti-Human, Clone: FP5A, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3564
Description
Detect Sodium/Iodine Symporter using this Anti-Sodium/Iodine Symporter Antibody, a.a. 625-643, clone FP5A validated for use in immunohistochemistry, immunohistochemistry (paraffin) and western blotting.Specifications
| Sodium/Iodine Symporter | |
| Monoclonal | |
| Unconjugated | |
| NM_000453.1 | |
| Mouse | |
| 100 μg | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| FP5A | |
| Liquid. Buffer = 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide. | |
| SLC5A5, +I, NIS, Na, -symporter), -cotransporter) | |
| Maltose Binding Protein fusion protein corresponding to amino acids 468-643 of human NIS (C-terminus). | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 6 months. | |
| IgG1 κ |
For Research Use Only