missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Channel NaB2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5358200UL
Description
Anti-Sodium Channel NaB2 Antibody detects level of Sodium Channel NaB2 and has been published and validated for use in immunohistochemistry and western blotting.Specifications
| Sodium Channel NaB2 | |
| Polyclonal | |
| Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, and 0.05 % sodium azide. Reconstitute with 200μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min). | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Rat |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| SCN2B; ORFNames=UNQ326/PRO386 | |
| Highly purified peptide corresponding to residues 197-215 of rat beta2 subunit of voltage-gated Na+ channels (Isom et al. 1995). Accession P54900. It is expected to work on human because the immunogen sequence is highly conserved (17/19 residues). Reacti | |
| 200 μL | |
| NM_004588.3 | |
| −20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only