missing translation for 'onlineSavingsMsg'
Learn More
Learn More
salt-inducible kinase 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human SNF1LK protein.
Supplier: Abnova Corporation H00150094B01P
Description
Sequence: MVIMSEFSADPAGQGQGQQKPLRVGFYDIERTLGKGNFAVVKLARHRVTKTQVAIKIIDKTRLDSSNLEKIYREVQLMKLLNHPHIIKLYQVMETKDMLYIVTEFAKNGEMFDYLTSNGHLSENEARKKFWQILSAVEYCHDHHIVHRDLKTENLLLDGNMDIKLADFGFGNFYKSGEPLSTWCGSPPYAAPEVFEGKEYEGPQLDIWSLGVVLYVLVCGSLPFDGPNLPTLRQRVLEGRFRIPFFMSQDCESLIRRMLVVDPARRITIAQIRQHRWMRAEPCLPGPACPAFSAHSYTSNLGDYDEQALGIMQTLGVDRQRTVESLQNSSYNHFAAIYYLLLERLKEYRNAQCARPGPARQPRPRSSDLSGLEVPQEGLSTDPFRPALLCPQPQTLVQSVLQAEMDCELQSSLQWPLFFPVDASCSGVFRPRPVSPSSLLDTAISEEARQGPGLEEEQDTQESLPSSTGRRHTLAEVSTRLSPLTAPCIVVSPSTTASPAEGTSSDSCLTFSASKSPAGLSGTPATQGLLGACSPVRLASPFLGSQSATPVLQAQGGLGGAVLLPVSFQEGRRASDTSLTQGLKAFRQQLRKTTRTKGFLGLNKIKGLARQVCQVPASRASRGGLSPFHAPAQSPGLHGGAAGSREGWSLLEEVLEQQRLLQLQHHPAAAPGCSQAPQPAPAPFVIAPCDGPGAAPLPSTLLTSGLPLLPPPLLQTGASPVASAAQLLDTHLHIGTGPTALPAVPPPRLARLAPGCEPLGLLQGDCEMEDLMPCSLGTFVLVQSpecifications
| salt-inducible kinase 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human SNF1LK protein. | |
| SIK1 | |
| MSK/SIK/SNF1LK | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_173354.2 | |
| SIK1 | |
| SNF1LK (AAH38504.1, 1 a.a. ∼ 783 a.a) full-length human protein. | |
| 50 μg | |
| Kinases | |
| 150094 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |