missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMRTe Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 06891
Description
Anti-SMRTe Antibody is a high quality Rabbit Polyclonal Antibody for the detection of SMRTe and has been validated in Chimmunoprecipitation, immunocytochemistry and western blotting.Specifications
| SMRTe | |
| Polyclonal | |
| 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% | |
| Rabbit | |
| Protein A chromatography | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| NCOR2; SMRTE; TRAC-1; CTG26; SMRTe; TNRC14; SMRT; TRAC1; SMAP270; SMRTE-tau; TRAC; N-CoR2 | |
| GST fusion-protein corresponding to residues 1146-1349 of human SMRTe | |
| 100 μg | |
| NM_006312 | |
| −20°C for 24 months | |
| IgG |
For Research Use Only