missing translation for 'onlineSavingsMsg'
Learn More
Learn More
solute carrier organic anion transporter family, member 2A1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLCO2A1.
Supplier: Abnova Corporation H00006578A01
Description
Sequence: FFFPRAMPIGAKRAPATADEARKLEEAKSRGSLVDFIKRFPCIFLRLLMNSpecifications
| solute carrier organic anion transporter family, member 2A1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SLCO2A1. | |
| SLCO2A1 | |
| OATP2A1/PGT/SLC21A2 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 6578 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_005630 | |
| SLCO2A1 | |
| SLCO2A1 (NP_005621, 276 a.a. ∼ 325 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |