missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Sialyl Lewis X Monoclonal Antibody , TCI America™
Anti-Sialyl Lewis X Monoclonal Antibody, TCI America™
Supplier: TCI America A28491VIAL
Specifications
| Sialyl Lewis X | |
| Monoclonal | |
| Mouse | |
| 1 Vial | |
| Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
| IgM κ |
| Immunohistochemistry, Flow Cytometry, ELISA, Western Blot, Inhibition Assays | |
| 2H5 | |
| A mixture of sialyl Lewis X-like determinants from human colon carcinoma cells | |
| Primary | |
| Purified |