Learn More
Anti-Sialyl Lewis A Monoclonal Antibody (1H4) , TCI America™
Anti-Sialyl Lewis A Monoclonal Antibody (1H4), TCI America™
Supplier:  TCI America A25841VIAL
Catalog No. A2584VIAL
Only null left
Description
Description
Specifications
Specifications
Sialyl Lewis A
Monoclonal
Mouse
1 Vial
Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing.
IgG3 κ
Immunohistochemistry, Flow Cytometry, ELISA, TLC-Immunostaining, Western Blot, Inhibition Assays
1H4
Immunized with human colon cancer cells (Colo201), and screened using sialyl Lewis A-active glycolipids
Primary
Purified
Product Suggestions
Product Suggestions