missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-SGPG(HNK-1) Monoclonal Antibody , TCI America™
Anti-SGPG(HNK-1) Monoclonal Antibody, TCI America™
Supplier: TCI America A27061VIAL
Specifications
| SGPG(HNK-1) | |
| Monoclonal | |
| Mouse | |
| 1 Vial | |
| Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
| IgM κ |
| TLC-Immunostaining, Flow Cytometry, ELISA, Immunohistochemistry, Western Blot | |
| NGR50 | |
| Purified sulfoglucuronosyl para- globoside (SGPG) adsorbed to Salmonella minnesota | |
| Primary | |
| Purified |