missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRP2 Mouse anti-Human, Clone: 80.8.6, MilliporeSigma™
Description
Anti-SFRP2 Antibody, clone 80.8.6, azide free is an antibody against SFRP2 for use in immunohistochemistry and inhibition.
Specifications
Specifications
| Antigen | SFRP2 |
| Applications | Immunohistochemistry, Inhibition Assays |
| Classification | Monoclonal |
| Clone | 80.8.6 |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal IgG2aκ in buffer containing PBS without preservatives. |
| Gene Accession No. | NP_003004 |
| Gene Symbols | SFRP2, FRP2, SARP1, FKSG12, UNQ361, PRO697 |
| Host Species | Mouse |
| Immunogen | Linear peptide corresponding to the NTR domain of human SFRP2. |
| Show More |
For Research Use Only