missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFN, Mouse, Clone: 1E6, Abnova™
Description
Sequence: MERASLIQKAKLAEQAERYEDMAAFMKGAVEKGEELSCEERNLLSVAYKNVVGGQRAAWRVLSSIEQKSNEEGSEEKGPEVREYREKAETELQGVCDTVLGLLDSHLIKEAGDAESRVFYLKMKGDYYRYLAEVATGDDKKRIIDSARSAYQEAMDISKKEMPPTNPIRLGLALNFSVFHYEIANSPEEAISLAKTTFDEAMADLHTLSEDSYKDSTLIMQLLRDNLTLWTADNAGEEGGEAPQEPQS
Specifications
Specifications
| Antigen | SFN |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 1E6 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full-length recombinant SFN. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SFN |
| Gene Accession No. | BC000329 |
| Gene Alias | YWHAS |
| Show More |