missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEMA4A, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SEMA4A.
Supplier: Abnova Corporation H00064218A01
Description
SEMA4A is a member of the semaphorin family of soluble and transmembrane proteins. Semaphorins are involved in guidance of axonal migration during neuronal development and in immune responses.[supplied by OMIM
Sequence: GGGQGPMPRVRYYAGDERRALSFFHQKGLQDFDTLLLSGDGNTLYVGAREAILALDIQDPGVPRLKNMIPWPASDRKKSECAFKKKSNETQCFNFIRVLVSpecifications
| SEMA4A | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SEMA4A. | |
| SEMA4A | |
| CORD10/FLJ12287/RP35/SEMAB/SEMB | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_022367 | |
| SEMA4A | |
| SEMA4A (NP_071762, 33 a.a. ∼ 132 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 64218 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |