missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sex comb on midleg homolog 1 (Drosophila), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SCMH1.
Supplier: Abnova Corporation H00022955A01
Description
Sequence: EKLCHNLRSDNLFGNQPFTQTHLSLTAIEYSHSHDRYLPGETFVLGNSLARSLEPHSDSMDSASNPTNLVSTSQRHRPLLSSCGLPPSTASAVRRLCSRSpecifications
| sex comb on midleg homolog 1 (Drosophila) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SCMH1. | |
| SCMH1 | |
| Scml3 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 22955 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_012236 | |
| SCMH1 | |
| SCMH1 (NP_036368, 404 a.a. ∼ 502 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |