Learn More
RAR-related orphan receptor B, Mouse, Clone: 1D1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RORB.
Supplier: Abnova Corporation H00006096M04
Description
The protein encoded by this gene is a member of the NR1 subfamily of nuclear hormone receptors. It is a DNA-binding protein that can bind as a monomer or as a homodimer to hormone response elements upstream of several genes to enhance the expression of those genes. The specific functions of this protein are not known, but it has been shown to interact with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation. [provided by RefSeq
Sequence: ETSGTYANGHVIDLPKSEGYYNVDSGQPSPDQSGLDMTGIKQIKQEPIYDLTSVPNLFTYSSFNNGQLAPGITMTEIDRIAQNIIKSHLSpecifications
| RAR-related orphan receptor B | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006914 | |
| RORB | |
| RORB (NP_008845, 136 a.a. ∼ 224 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1D1 | |
| Mouse monoclonal antibody raised against a partial recombinant RORB. | |
| RORB | |
| NR1F2/ROR-BETA/RZR-BETA/RZRB/bA133M9.1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 6096 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 λ |