missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodopsin Mouse anti-Vertebrates, Clone: Rho 1D4, MilliporeSigma™
Description
Anti-Rhodopsin Antibody, C-terminus, last 9 amino acids, clone Rho 1D4 detects level of Rhodopsin and has been published and validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
Specifications
Specifications
| Antigen | Rhodopsin |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | Rho 1D4 |
| Conjugate | Unconjugated |
| Formulation | Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative. |
| Gene Accession No. | NM_000539.2 |
| Gene Symbols | RHO, rhodopsin, Opsin-2, OPN2, Rhodopsin, RP4, MGC138311, MGC138309 |
| Host Species | Mouse |
| Immunogen | Bovine rhodopsin |
| Show More |
For Research Use Only