missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Rabbit IgG, peroxidase-linked species-specific whole antibody (from donkey) Secondary Antibody, Cytiva
Description
- Host Species: Donkey
- Isotype: IgG
- Species Reactivity: Rb
- Immunogen: Rabbit IgG
- Applications: Protein Blotting, ELISA, Immunocytochemistry
Specifications
Specifications
| Antigen | IgG |
| Classification | Monoclonal |
| Conjugate | HRP |
| Host Species | Donkey |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mL |
| Primary or Secondary | Secondary |
| Isotype | IgG |