missing translation for 'onlineSavingsMsg'
Learn More

anti-Rabbit IgG, peroxidase-linked species-specific whole antibody (from donkey) Secondary Antibody, Cytiva

Catalog No. 45001276
Change view
Click to view available options
Quantity:
0.1 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001276 0.1 mL
45000682 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45001276 Supplier Cytiva Supplier No. NA934100UL
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A highly species-specific anti-Rabbit IgG. Cytiva Lifescience™ anti-Rabbit IgG, peroxidase-linked species-specific whole antibody (from donkey) Secondary Antibody is used with Amersham™ ECL Western Blotting detection reagents.

  • Host Species: Donkey
  • Isotype: IgG
  • Species Reactivity: Rb
  • Immunogen: Rabbit IgG
  • Applications: Protein Blotting, ELISA, Immunocytochemistry

Specifications

Antigen IgG
Classification Monoclonal
Conjugate HRP
Host Species Donkey
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Secondary
Isotype IgG