Learn More
Invitrogen™ F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555 DFS Item
F(ab')2-Goat Polyclonal Secondary Antibody
Supplier:  Invitrogen™ A21430
Catalog No. A21430
Only null left
Explore available promotions
Description
Description
Product will be shipped at Room Temperature.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Rabbit IgG (H+L) Cross-Adsorbed
Polyclonal
Alexa Fluor 555
Goat
Purified
RUO
Rabbit
Secondary Antibody
IgG
Western Blot, Immunocytochemistry
2 mg/mL
PBS with 5mM sodium azide; pH 7.5
Gamma Immunoglobins Heavy and Light chains.
500 μg
Secondary
4°C, store in dark
Liquid
Product Suggestions
Product Suggestions