missing translation for 'onlineSavingsMsg'
Learn More

RA-induced protein 17 Mouse anti-Human, Clone: 5H5.1, MilliporeSigma™

Catalog No. MABN250MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN250MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN250MI Supplier MilliporeSigma Supplier No. MABN250
Only null left

Mouse Monoclonal Antibody

Detect Retinoic acid-induced protein 17 using this mouse monoclonal antibody, Anti-RA-induced protein 17 Antibody, clone 5H5.1 validated for use in western blotting and immunohistochemistry.

Specifications

Antigen RA-induced protein 17
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5H5.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_065071
Gene Symbols ZMIZ1, KIAA1224, RAI17, ZIMP10
Host Species Mouse
Immunogen Recombinant protein corresponding to human RA-induced protein 17.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only