missing translation for 'onlineSavingsMsg'
Learn More

Puromycin Mouse anti-Human, All, Clone: 12D10, MilliporeSigma™

Catalog No. MABE343MI
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE343MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE343MI Supplier MilliporeSigma Supplier No. MABE343

Mouse Monoclonal Antibody

Anti-Puromycin antibody, clone 12D10, detects puromycin incorporated into protein. Monoclonal antibodies to puromycin may be used with standard immunochemical methods.

Specifications

Antigen Puromycin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 12D10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Puromycin from Streptomyces alboniger
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species All species, Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only