missing translation for 'onlineSavingsMsg'
Learn More

Prostaglandin E Receptor EP2 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB9472MI
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9472MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9472MI Supplier MilliporeSigma Supplier No. AB9472
Only null left

Rabbit Polyclonal Antibody

Detect Prostaglandin E Receptor EP2 using this Anti-Prostaglandin E Receptor EP2 Antibody validated for use in immunohistochemistry (paraffin).

Specifications

Antigen Prostaglandin E Receptor EP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide.
Gene Symbols PTGER2; EP2
Host Species Rabbit
Immunogen Synthetic peptide from the 3rd extracellular domain of human Prostaglandin E Receptor EP2.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000956.2
Target Species Human
Content And Storage −70°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only