missing translation for 'onlineSavingsMsg'
Learn More
Learn More
protein kinase, interferon-inducible double stranded RNA dependent activator, Mouse, Clone: 1B9-1A7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant PRKRA.
Supplier: Abnova Corporation H00008575M01
Description
This gene encodes a protein kinase activated by double-stranded RNA which mediates the effects of interferon in response to viral infection. Mutations in this gene have been associated with dystonia. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: MSQSRHRAEAPPLEREDSGTFSLGKMITAKPGKTPIQVLHEYGMKTKNIPVYECERSDVQIHVPTFTFRVTVGDITCTGEGTSKKLAKHRAAEAAINILKANASICFAVPDPLMPDPSKQPKNQLNPIGSLQELAIHHGWRLPEYTLSQEGGPAHKREYTTICRLESFMETGKGASKKQAKRNAAEKFLAKFSNISPENHISLTNVVGHSLGCTWHSLRNSPGEKINLLKRSLLSIPNTDYIQLLSEIAKEQGFNITYLDIDELSANGQYQCLAELSTSPITVCHGSGISCGNAQSDAAHNALQYLKIIAERKSpecifications
| protein kinase, interferon-inducible double stranded RNA dependent activator | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC009470 | |
| PRKRA | |
| PRKRA (AAH09470, 1 a.a. ∼ 313 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| 1B9-1A7 | |
| Mouse monoclonal antibody raised against a full length recombinant PRKRA. | |
| PRKRA | |
| DYT16/HSD14/PACT/RAX | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 8575 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |