Learn More
PRDM1 (BLIMP-1) Rat anti-Human, Mouse, Clone: 6D3, MilliporeSigma™
Rat Monoclonal Antibody
Supplier:  MilliporeSigma 051570
Catalog No. 05-157-0MI
Only null left
Explore available promotions
Description
Description
Use Anti-PRDM1 (BLIMP-1) Antibody, clone 6D3 (Rat Monoclonal Antibody) validated in western blot, ELISA, immunohistochemistry, FC to detect PRDM1 (BLIMP-1) also known as PR-domain zinc finger protein 1.
Specifications
Specifications
PRDM1 (BLIMP-1)
Monoclonal
Unconjugated
NP_031574
Rat
Protein G
Primary
2-8°C for one year
IgG2a κ
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
6D3
Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
BLIMP1, PRDM1, PRDI-BF1, BLIMP-1
Recombinant protein corresponding to mouse BLIMP-1.
100 μg
Human, Mouse
Purified
Product Suggestions
Product Suggestions