missing translation for 'onlineSavingsMsg'
Learn More

PPARGC1A, Mouse, Clone: 1E11, Abnova™

Mouse monoclonal antibody raised against a partial recombinant PPARGC1A.

Supplier:  Abnova Corporation H00010891M05

Catalog No. 89-002-780


Only null left
Explore available promotions

Description

Description

The protein encoded by this gene is a transcriptional coactivator that regulates the genes involved in energy metabolism. This protein interacts with PPARgamma, which permits the interaction of this protein with multiple transcription factors. This protein can interact with, and regulate the activities of, cAMP response element binding protein (CREB) and nuclear respiratory factors (NRFs). It provides a direct link between external physiological stimuli and the regulation of mitochondrial biogenesis, and is a major factor that regulates muscle fiber type determination. This protein may be also involved in controlling blood pressure, regulating cellular cholesterol homoeostasis, and the development of obesity. [provided by RefSeq

Sequence: TRTELRDRFEVFGEIEECTVNLRDDGDSYGFITYRYTCDAFAALENGYTLRRSNETDFELYFCGRKQFFKSNYADLDSNSDDFDPASTKSKYDSLDFDSLLKEAQRSLRR
Specifications

Specifications

PPARGC1A,
Monoclonal
Unconjugated
In 1X PBS, pH 7.4
NM_013261
PPARGC1A
PPARGC1A (NP_037393, 689 a.a. ∼ 798 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
10891
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG2a κ
ELISA, Western Blot
1E11
Mouse monoclonal antibody raised against a partial recombinant PPARGC1A.
PPARGC1A
LEM6/PGC-1(alpha)/PGC-1v/PGC1/PGC1A/PPARGC1
Mouse
100 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions