missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP5 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 071225
Description
Detect PP5 using this Anti-PP5 Antibody validated for use in western blot, immunohistochemistry (paraffin).Specifications
| PP5 | |
| Polyclonal | |
| Purified rabbit polyclonal IgG in phosphate buffered saline with 0.08% sodium azide | |
| Rabbit | |
| 100 μg | |
| NM_006247.2 | |
| −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| PPP5C; PPP5; FLJ36922; PP-T; PP5; PPT; EC 3.1.3.16 | |
| Synthetic peptide derived from the human protein phosphatase 5 protein. | |
| Primary | |
| Human, Mouse | |
| Purified |
For Research Use Only