missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Channel MaxiK Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB522850UL
Description
Anti-Potassium Channel MaxiK Antibody, calcium activated, aa1097-1196 detects level of Potassium Channel MaxiK and has been published and validated for use in western blotting.Specifications
| Potassium Channel MaxiK | |
| Polyclonal | |
| Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, 5% sucrose as a stabilizer and 0.025% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody prep | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Rat |
| Western Blot | |
| Unconjugated | |
| KCNMA1; OTTHUMP00000064161; OTTHUMP00000060198; KCNMA; K; OTTHUMP00000064158; MaxiK; VCAlpha); SLO-ALPHA; MGC71881; OTTHUMP00000064157; SAKCA; OTTHUMP00000064156; OTTHUMP00000064164; Slo1; BKTM; OTTHUMP00000064165; hSlo; mSLO1; OTTHUMP00000064154; Slo-alpha; SLO; OTTHUMP000000 | |
| A purified fusion protein (GST)1 and a C-terminal part (residues 1097-1196) of mouse Slo (mSlo) alpha subunit of BKCa channel. | |
| 50 μL | |
| NM_001014797.1; NM_002247.2 | |
| −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only